Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A2B1 Monoclonal Antibody (C.492.9), Invitrogen™

Catalog No. PIMA515106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515106 Supplier Thermo Scientific Supplier No. MA515106

Mouse Monoclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other proteins.

This gene belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. This gene has been described to generate two alternatively spliced transcript variants which encode different isoforms. [provided by RefSeq, Jul 2008]
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP A2B1
Applications Western Blot
Classification Monoclonal
Clone C.492.9
Conjugate Unconjugated
Formulation 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
Gene HNRNPA2B1
Gene Accession No. P22626
Gene Alias 9130414A06Rik; DKFZp779B0244; EMBL:BAF79676.1}; FLJ22720; heterogeneous nuclear ribonucleoprotein A2; heterogeneous nuclear ribonucleoprotein A2/B1; heterogeneous nuclear ribonucleoprotein B0a; heterogeneous nuclear ribonucleoprotein B0b; heterogeneous nuclear ribonucleoprotein B1; heterogeneous nuclear ribonucleoproteins A2/B1; heterogeneous nuclear ribonucleoproteins A2/B1 {ECO:0000312; heterogenous nuclear ribonucleoprotein A2/B1; Hnrnp; hnRNP A2 / hnRNP B1; hnRNP A2/B1; hnrnp-A; HNRNPA2; HNRNPA2B1; hnrnpa2b1 {ECO:0000250; HNRNPB1; HNRPA2; HNRPA2B1; HNRPB1; hypothetical protein; IBMPFD2; nuclear ribonucleoprotein particle A2 protein; OTTHUMP00000202465; OTTHUMP00000202466; RNPA2; SNRPB1; UniProtKB:P22626}
Gene Symbols HNRNPA2B1
Host Species Mouse
Immunogen Human hnRNP B1 protein
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3181
Target Species Human, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.