Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A2B1 Mouse anti-Human, Mouse, Clone: 12B12.2, MilliporeSigma™

Catalog No. MABF1098MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1098MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1098MI Supplier MilliporeSigma Supplier No. MABF1098

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

This mouse monoclonal Anti-hnRNP A2B1, clone 12B12.2 Antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP A2B1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 12B12.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols HNRNPA2B1
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human heterogeneous nuclear ribonucleoproteins A2/B1.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_112533
Test Specificity Clone 12B12.2 specifically detects heterogeneous nuclear ribonucleoproteins A2/B1. it targets an epitope within 15 amino acids from the N-terminal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.