Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hnRNP A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116943
Description
Positive Samples: U-251MG Immunogen sequence: VDGRVVEPKR AVSREDSVKP GAHLTVKKIF VGGIKEDTEE YNLRDYFEKY GKIETIEVME DRQSGKKRGF AFVTFDDHDT VDKIVVQKYH TINGHNCEVK K.
HNRPA3 contains 2 RRM (RNA recognition motif) domains and plays a role in cytoplasmic trafficking of RNA. It binds to the cis-acting response element, A2RE and may be involved in pre-mRNA splicing.
Specifications
hnRNP A3 | |
Polyclonal | |
Unconjugated | |
HNRNPA3 | |
2410013L13Rik; 2610209F03Rik; 2610510D13Rik; D10S102; FBRNP; Heterogeneous nuclear ribonucleoprotein A3; hnRNP A3; Hnrnpa3; HNRPA3; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein A3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
220988, 229279, 362152 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2.47 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P51991, Q6URK4, Q8BG05 | |
HNRNPA3 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human HNRNPA3 (NP_0013171781). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction