Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541961 Supplier Invitrogen™ Supplier No. PA541961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MEVKPPPGRP QPDSGRRRRR RGEEGHDPKE PEQLRKLFIG GLSFETTDDS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

HNRPA3 contains 2 RRM (RNA recognition motif) domains and plays a role in cytoplasmic trafficking of RNA. It binds to the cis-acting response element, A2RE and may be involved in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HNRNPA3
Gene Accession No. P51991
Gene Alias 2410013L13Rik; 2610209F03Rik; 2610510D13Rik; D10S102; FBRNP; Heterogeneous nuclear ribonucleoprotein A3; hnRNP A3; Hnrnpa3; HNRPA3; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein A3
Gene Symbols HNRNPA3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HNRPA3 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220988
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.