Learn More
Invitrogen™ hnRNP AB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113701
Description
Immunogen sequence: GAAAGAGGAT AAPPSGNQNG AEGDQINASK NEEDAGKMFV GGLSWDTSKK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 86%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 93%.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are produced by RNA polymerase II and are components of the heterogeneous nuclear RNA (hnRNA) complexes. They are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene, which binds to one of the components of the multiprotein editosome complex, has two repeats of quasi-RRM (RNA recognition motif) domains that bind to RNAs. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| hnRNP AB | |
| Polyclonal | |
| Unconjugated | |
| HNRNPAB | |
| 3010025C11Rik; A1F-C1; ABBP1; ABBP-1; apobec-1 binding protein 1; APOBEC1-binding protein 1; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1-binding protein 1; CArG-binding factor-A; Cbf-a; Cgbfa; Heterogeneous nuclear ribonucleoprotein A/B; hnRNP A/B; hnRNP type A/B protein; HNRNPAB; Hnrpab; S1 protein D2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3182 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99729 | |
| HNRNPAB | |
| Synthetic peptide directed towards the N terminal region of human HNRPAB (aa 34-83). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.