Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP AB Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | hnRNP AB |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
hnRNP AB Polyclonal specifically detects hnRNP AB in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
hnRNP AB | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
catalytic polypeptide 1-binding protein 1, heterogeneous nuclear ribonucleoprotein A/B, hnRNP A/B, hnRNP type A/B protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of human hnRNP AB (NP_004490.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS with 50% glycerol, pH7.3. | |
3182 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title