Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584048 Supplier Invitrogen™ Supplier No. PA584048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEMYGSVTEH PSPSPLLSSS Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 95%.

This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene can act as a tetramer and is involved in the assembly of 40S hnRNP particles. Multiple transcript variants encoding at least two different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HNRNPC
Gene Accession No. P07910
Gene Alias AL022939; C1; C2; D14Wsu171e; heterogeneous nuclear ribonucleoprotein C; heterogeneous nuclear ribonucleoprotein C (C1/C2); heterogeneous nuclear ribonucleoprotein C1; heterogeneous nuclear ribonucleoprotein C2; heterogeneous nuclear ribonucleoproteins C1/C2; heterogeneous ribonucleoprotein C1/C2; HNRNP; hnRNP C; hnRNP C protein C-terminal fragment (158 AA); hnRNP C1 / hnRNP C2; hnRNP C1/C2; hnRNP C2; hnRNP core protein C; Hnrnpc; hnrnp-C; hnRNPC1; hnRNPC2; HNRPC; Hnrpc1; Hnrpc2; nuclear ribonucleoprotein particle C1 protein; nuclear ribonucleoprotein particle C2 protein; snoRNA MBI-122; SNRPC
Gene Symbols HNRNPC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HNRNPC. Recombinant protein control fragment (Product #RP-101144).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3183
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.