Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP C1/C2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599503
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599503 Supplier Invitrogen™ Supplier No. PA599503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total hnRNP C1/C2.

The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. hnRNP C1 and C2 are encoded by one gene; the two alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP C1/C2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HNRNPC
Gene Accession No. G3V9R8, P07910, Q9Z204
Gene Alias AL022939; bZ1G18.4; C1; C2; D14Wsu171e; fb57h12; fi16b11; heterogeneous nuclear ribonucleoprotein C; heterogeneous nuclear ribonucleoprotein C (C1/C2); heterogeneous nuclear ribonucleoprotein C1; heterogeneous nuclear ribonucleoprotein C2; heterogeneous nuclear ribonucleoproteins C1/C2; heterogeneous nuclear ribonucleoproteins C1/C2; heterogeneous nuclear ribonucleoprotein C; heterogeneous ribonucleoprotein C1/C2; HNRNP; hnRNP C; hnRNP C protein C-terminal fragment (158 AA); hnRNP C1 / hnRNP C2; hnRNP C1/C2; hnRNP C2; hnRNP core protein C; HNRNPC; hnrnp-C; hnRNPC1; hnRNPC2; HNRPC; Hnrpc1; Hnrpc2; hypothetical protein LOC406352; nuclear ribonucleoprotein particle C1 protein; nuclear ribonucleoprotein particle C2 protein; snoRNA MBI-122; SNRPC; wu:fb57h12; wu:fi16b11; zgc:55701
Gene Symbols HNRNPC
Host Species Rabbit
Immunogen A synthesized peptide derived from human HNRNPC(Accession P07910), corresponding to amino acid residues E236-D286.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15381, 290046, 3183
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.