Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599155 Supplier Thermo Scientific Supplier No. PA599155
Add to Cart
Add to Cart

hnRNP D Polyclonal Antibody

hnRNP D Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HNRNPD
Gene Accession No. Q14103, Q60668, Q9JJ54
Gene Alias ARE-binding protein AUFI, type A; Auf1; AUF1A; AU-rich element RNA binding protein 1, 37kDa; AU-rich element RNA-binding factor 1; AU-rich element RNA-binding protein 1; heterogeneous nuclear ribonucleoprotein D; heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa); heterogeneous nuclear ribonucleoprotein D0; hnRNP D0; HNRNPD; hnRNPD0; Hnrpd; P37; RNA binding protein p45AUF1
Gene Symbols HNRNPD
Host Species Rabbit
Immunogen A synthesized peptide derived from human HNRNPD(Accession Q14103), corresponding to amino acid residues G59-T109.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11991, 3184, 79256
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.