Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hnRNP DL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563354
Description
Immunogen sequence: DSSVTMEDMN EYSNIEEFAE GSKINASKNQ QDDGKM Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
hnRNP DL belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two RRM domains that bind to RNAs. Three alternatively spliced transcript variants have been described for this gene. One of the variants is probably not translated because the transcript is a candidate for nonsense-mediated mRNA decay. The protein isoforms encoded by this gene are similar to its family member HNRPD.
Specifications
hnRNP DL | |
Polyclonal | |
Unconjugated | |
HNRNPDL | |
A+U-rich element RNA binding factor; AA407431; AA959857; AU-rich element RNA-binding factor; D5Ertd650e; D5Wsu145e; heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa); heterogeneous nuclear ribonucleoprotein D like; heterogeneous nuclear ribonucleoprotein D-like; heterogenous ribonucleoprotein D-like; HNRNP; hnRNP DL; hnRNP D-like; HNRNPD; Hnrnpdl; hnRNP-DL; Hnrnpdl1; HNRPDL; hnRPD-like protein; JKT41-binding protein; JKTBP; JKTBP2; laAUF1; LGMD1G; LOC534770 protein; protein laAUF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14979 | |
HNRNPDL | |
Recombinant protein corresponding to Human hnRNP DL. Recombinant protein control fragment (Product #RP-103272). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction