Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP DL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103885 Supplier Invitrogen™ Supplier No. PA5103885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HNRPDL.

hnRNP DL belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two RRM domains that bind to RNAs. Three alternatively spliced transcript variants have been described for this gene. One of the variants is probably not translated because the transcript is a candidate for nonsense-mediated mRNA decay. The protein isoforms encoded by this gene are similar to its family member HNRPD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP DL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HNRNPDL
Gene Accession No. O14979, Q3SWU3, Q9Z130
Gene Alias A+U-rich element RNA binding factor; AA407431; AA959857; AU-rich element RNA-binding factor; D5Ertd650e; D5Wsu145e; heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa); heterogeneous nuclear ribonucleoprotein D like; heterogeneous nuclear ribonucleoprotein D-like; heterogenous ribonucleoprotein D-like; HNRNP; hnRNP DL; hnRNP D-like; HNRNPD; Hnrnpdl; hnRNP-DL; Hnrnpdl1; HNRPDL; hnRPD-like protein; JKT41-binding protein; JKTBP; JKTBP2; laAUF1; LGMD1G; LOC534770 protein; protein laAUF1
Gene Symbols HNRNPDL
Host Species Rabbit
Immunogen A synthesized peptide derived from human HNRNPDL(Accession O14979), corresponding to amino acid residues G385-Y420.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305178, 50926, 9987
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.