Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP F Antibody (OTI5F5), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270907MFV450
Description
hnRNP F Monoclonal antibody specifically detects hnRNP F in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
hnRNP F | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI5F5 | |
Flow Cytometry | |
heterogeneous nuclear ribonucleoprotein F, hnRNP F, HNRPFHnRNP F protein, mcs94-1, MGC110997, Nucleolin-like protein mcs94-1, OK/SW-cl.23 | |
Full length human recombinant protein of human HNRNPF (NP_004957) produced in HEK293T cell. | |
0.1 mL | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
3185 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction