Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP F Monoclonal Antibody (3H4)
GREENER_CHOICE

Catalog No. PIMA134551
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA134551 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA134551 Supplier Invitrogen™ Supplier No. MA134551
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-34551 detects HNRNP F from human samples.

This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins. The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA. These proteins are associated with pre-mRNAs in the nucleus and regulate alternative splicing, polyadenylation, and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs which have guanosine-rich sequences. This protein is very similar to the family member hnRPH. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP F
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3H4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Hnrnpf
Gene Accession No. P52597
Gene Alias 4833420I20Rik; AA407306; heterogeneous nuclear ribonucleoprotein F; Heterogeneous nuclear ribonucleoprotein F, N-terminally processed; hnRNP F; HnRNP F protein; hnRNP H; Hnrnpf; HNRNPH1; Hnrpf; mcs94 1; mcs94-1; Nucleolin-like protein mcs94-1; OK/SW-cl.23; ribonucleoprotein F; ROF
Gene Symbols Hnrnpf
Host Species Mouse
Immunogen Full length hnRNP F protein (human).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3185
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.