Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hnRNP F Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549813
Description
The antibody detects endogenous levels of total hnRNP F protein.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins. The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA. These proteins are associated with pre-mRNAs in the nucleus and regulate alternative splicing, polyadenylation, and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs which have guanosine-rich sequences. This protein is very similar to the family member hnRPH. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
hnRNP F | |
Polyclonal | |
Unconjugated | |
Hnrnpf | |
4833420I20Rik; AA407306; heterogeneous nuclear ribonucleoprotein F; Heterogeneous nuclear ribonucleoprotein F, N-terminally processed; hnRNP F; HnRNP F protein; hnRNP H; Hnrnpf; HNRNPH1; Hnrpf; mcs94 1; mcs94-1; Nucleolin-like protein mcs94-1; OK/SW-cl.23; ribonucleoprotein F; ROF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3185, 64200, 98758 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P52597, Q794E4, Q9Z2X1 | |
Hnrnpf | |
Synthesized peptide derived from N-terminal of human hnRNP F. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction