Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP F Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303421100UL
Description
hnRNP F Polyclonal specifically detects hnRNP F in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
hnRNP F | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
heterogeneous nuclear ribonucleoprotein F, hnRNP F, HNRPFHnRNP F protein, mcs94-1, MGC110997, Nucleolin-like protein mcs94-1, OK/SW-cl.23 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human hnRNP F (NP_001091678.1). | |
100 μL | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
3185 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction