Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP G Rabbit anti-Human, Monkey, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30355020UL
Description
hnRNP G Polyclonal specifically detects hnRNP G in Human, Monkey samples. It is validated for Western Blot.Specifications
hnRNP G | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
27316 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
Glycoprotein p43, heterogeneous nuclear ribonucleoprotein G, hnRNP G, hnRNP-G, HNRPG, RBMXP1, RBMXRT, RNA binding motif protein, X chromosome, RNA binding motif protein, X-linked, RNA-binding motif protein, X chromosome, RNMX | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-95 of human hnRNP G (NP_002130.2). | |
20 μL | |
Primary | |
Human, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction