Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP H Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100385FR
Description
hnRNP H Polyclonal antibody specifically detects hnRNP H in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
hnRNP H | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
DKFZp686A15170, heterogeneous nuclear ribonucleoprotein H, heterogeneous nuclear ribonucleoprotein H1 (H), hnRNP H, HNRPH, HNRPH1hnRNPH | |
The immunogen recognized by this antibody maps to a region between residue 400 and the C-terminus (residue 448) of human Heterogeneous Nuclear Ribonucleoprotein H1 using the numbering given in entry NP_005511.1 (GeneID 3187). | |
0.1 mL | |
Cellular Markers, Translation Control | |
3187 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction