Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hnRNP H1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585159
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Positive Control: Jurkat, Raji, NCI-H929. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of a subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA. These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some may shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNA and is very similar to the family member HNRPF. This gene may be associated with hereditary lymphedema type I. Alternatively spliced transcript variants have been described.
Specifications
hnRNP H1 | |
Polyclonal | |
Unconjugated | |
Hnrnph1 | |
AI642080; E430005G16Rik; Heterogeneous nuclear ribonucleoprotein H; Heterogeneous nuclear ribonucleoprotein H, N-terminally processed; heterogeneous nuclear ribonucleoprotein H1; heterogeneous nuclear ribonucleoprotein H1 (H); hnRNP H; Hnrnph; HNRNPH1; HNRPH; HNRPH1; Ratsg1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3187 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.38 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P31943 | |
Hnrnph1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human hnRNP H. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction