Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP H2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551566 Supplier Invitrogen™ Supplier No. PA551566
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGRVTGEADV EFATHEDAVA AMAKDKANMQ HRYVELFLNS TAGTSGGAYD HSYVELFLNS TAGASGGAYG SQMMGGMGLS NQSSYGGPAS QQLSGGYGGG YGGQSSMSGY DQVLQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that binds to RNAs. It is very similar to the family member HNRPH1. This gene is thought to be involved in Fabray disease and X-linked agammaglobulinemia phenotype. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP H2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HNRNPH2
Gene Accession No. P55795
Gene Alias DXHXS1271E; Ftp3; Ftp-3; H'; Heterogeneous nuclear ribonucleoprotein H'; heterogeneous nuclear ribonucleoprotein H2; heterogeneous nuclear ribonucleoprotein H2 (H'); Heterogeneous nuclear ribonucleoprotein H2, N-terminally processed; heterogeneous nuclear ribonucleoprotein H-prime; HNRNP; hnRNP H'; hnRNP H2; hnRNPH'; Hnrnph2; HNRPH'; HNRPH2
Gene Symbols HNRNPH2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human HNRNPH2 (Product #RP-91891)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3188
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.