Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP H3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541837 Supplier Invitrogen™ Supplier No. PA541837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA5-41837 detects HNRPH3 in WB and IHC applications. Peptide sequence: DYQGRSTGEA FVQFASKEIA ENALGKHKER IGHRYIEIFR SSRSEIKGFY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

HNRPH3 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two repeats of quasi-RRM domains that bind to RNAs. It is localized in nuclear bodies of the nucleus. This protein is involved in the splicing process and it also participates in early heat shock-induced splicing arrest by transiently leaving the hnRNP complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP H3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HNRNPH3
Gene Accession No. P31942
Gene Alias 2H9; AA693301; AI666703; heterogeneous nuclear ribonucleoprotein 2H9; Heterogeneous nuclear ribonucleoprotein H3; heterogeneous nuclear ribonucleoprotein H3 (2H9); hnRNP 2H9; hnRNP H3; HNRNPH3; HNRPH3; Unknown (protein for MGC:136953); zgc:136953
Gene Symbols HNRNPH3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HNRPH3 (aa 51-100).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3189
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.