Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP K Antibody (N10 P2D3*G2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2504240.025ML
Description
hnRNP K Monoclonal antibody specifically detects hnRNP K in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
hnRNP K | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
dC-stretch binding protein, FLJ41122, heterogeneous nuclear ribonucleoprotein K, hnRNP K, transformation upregulated nuclear protein, Transformation up-regulated nuclear protein, TUNPHNRPKCSBP | |
Ovalbumin-conjugated synthetic peptide SVKQYSGKFF (C-terminal sequence) | |
0.025 mL | |
Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
3190 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Immunohistochemistry | |
N10 P2D3*G2 | |
Unconjugated | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction