Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP K Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen hnRNP K
Concentration 0.5 mg/mL
Dilution Western Blot 2-5 ug/ml. Use reported by customer review, Immunohistochemistry, Immunohistochemistry-Paraffin 5-10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224531SS
SDP
View Documents
Novus Biologicals
NBP224531SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP224531
SDP
View Documents
Novus Biologicals
NBP224531
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

hnRNP K Polyclonal specifically detects hnRNP K in Human, Mouse, Rat, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

hnRNP K
Western Blot 2-5 ug/ml. Use reported by customer review, Immunohistochemistry, Immunohistochemistry-Paraffin 5-10 ug/ml
Polyclonal
Rabbit
Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
3190
A portion of amino acid 1 to 50 of human hnRNPK was used as immunogen for the antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat, Opossum, Primate
dC-stretch binding protein, FLJ41122, heterogeneous nuclear ribonucleoprotein K, hnRNP K, transformation upregulated nuclear protein, Transformation up-regulated nuclear protein, TUNPHNRPKCSBP
HNRNPK
IgG
Affinity Purified
The amino acid sequence used as immunogen for the Importin-8 antibody is 100% homologous in human and chimpanzee and monkey, 97% incow and dog, 92% in mouse, and 86% homologous inchicken and opossum.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.