Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP K/J Rabbit anti-Human, Mouse, Rat, Clone: S07-5H0, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | hnRNP K/J |
---|---|
Clone | S07-5H0 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunofluorescence 10 μg/mL |
Applications | Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Monoclonal |
Description
hnRNP K/J Monoclonal antibody specifically detects hnRNP K/J in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
hnRNP K/J | |
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunofluorescence 10 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
dC-stretch binding protein, FLJ41122, heterogeneous nuclear ribonucleoprotein K, hnRNP K, transformation upregulated nuclear protein, Transformation up-regulated nuclear protein, TUNPHNRPKCSBP | |
A synthetic peptide of human hnRNP K/J (Uniprot#: P61978) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S07-5H0 | |
Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
3190 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title