Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP L Antibody (4D11), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1206106H
Description
hnRNP L Monoclonal specifically detects hnRNP L in Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
hnRNP L | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
HNRNPL | |
Human hnRNP proteins from HeLa cells purified by affinity chromatography on ssDNA agarose. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
4D11 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL | |
Mouse | |
Protein G purified | |
Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
3191 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction