Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP L Antibody (OTI2E4), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270908AF594
Description
hnRNP L Monoclonal specifically detects hnRNP L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
hnRNP L | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
HNRNPL | |
Recombinant protein fragment corresponding to amino acids 270-502 of human hnRNP L (NP_001524) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2E4 | |
Western Blot, Immunohistochemistry | |
FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL | |
Mouse | |
0.1 mL | |
3191 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction