Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP L Monoclonal Antibody (4D11)
GREENER_CHOICE

Catalog No. PIMA126745
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA126745 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA126745 Supplier Invitrogen™ Supplier No. MA126745
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: MDBK. A band of ~65 kDa can be detected.

Heterogeneous nuclear RNAs (hnRNAs) which include mRNA precursors and mature mRNAs are associated with specific proteins to form heterogenous ribonucleoprotein (hnRNP) complexes. Heterogeneous nuclear ribonucleoprotein L is among the proteins that are stably associated with hnRNP complexes and along with other hnRNP proteins is likely to play a major role in the formation, packaging, processing, and function of mRNA. Heterogeneous nuclear ribonucleoprotein L is present in the nucleoplasm as part of the HNRP complex. HNRP proteins have also been identified outside of the nucleoplasm. Exchange of hnRNP for mRNA-binding proteins accompanies transport of mRNA from the nucleus to the cytoplasm. Since HNRP proteins have been shown to shuttle between the nucleus and the cytoplasm, it is possible that they also have cytoplasmic functions. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP L
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4D11
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene HNRNPL
Gene Accession No. P14866, Q8R081
Gene Alias C79783; D830027H13Rik; Fblim1; FLJ35509; Heterogeneous nuclear ribonucleoprotein L; heterogenous nuclear ribonucleoprotein L; hnRNP L; Hnrnpl; hnRNP-L; Hnrpl; P/OKcl.14
Gene Symbols HNRNPL
Host Species Mouse
Immunogen HnRNP proteins.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101750874, 15388, 3191, 504520
Target Species Amphibian, Bovine, Chicken, Human, Mouse, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.