Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597050 Supplier Invitrogen™ Supplier No. PA597050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TWDYTNPNLS GQGDPGSNPN KRQRQPPLLG DHPAEYGGPH GGYHSHYHDE GYGPPPPHYE GRRMGPPVGG HRRGPSRYGP QYGHPPPPPP PPEYGPHADS PVLMVYGLDQ SKMNCDRVFN VFCLYGNVEK VKFMKSKPGA AMVEMADGYA VDRAITHLNN NFMFGQKLNV CVSKQPAIMP GQSYGLEDGS CSYKDFSESR NNRFSTPEQA AKNRIQHPSN VLHFFNAPLE VTEENFFEIC DELGVKRPSS VKVFSGKSER SSSGLLEWES KSDALETLGF LNHYQMKNPN GPYPYTLKLC FSTAQHAS; Positive Samples: U-87MG, HeLa, HepG2, A-549, HT-29, Mouse ovary, Rat lung, Rat brain; Cellular Location: Cytoplasm, Nucleus, nucleoplasm.

Heterogeneous nuclear RNAs (hnRNAs) which include mRNA precursors and mature mRNAs are associated with specific proteins to form heterogenous ribonucleoprotein (hnRNP) complexes. Heterogeneous nuclear ribonucleoprotein L is among the proteins that are stably associated with hnRNP complexes and along with other hnRNP proteins is likely to play a major role in the formation, packaging, processing, and function of mRNA. Heterogeneous nuclear ribonucleoprotein L is present in the nucleoplasm as part of the HNRP complex. HNRP proteins have also been identified outside of the nucleoplasm. Exchange of hnRNP for mRNA-binding proteins accompanies transport of mRNA from the nucleus to the cytoplasm. Since HNRP proteins have been shown to shuttle between the nucleus and the cytoplasm, it is possible that they also have cytoplasmic functions. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP L
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HNRNPL
Gene Accession No. F1LQ48, P14866, Q8R081
Gene Alias C79783; D830027H13Rik; Fblim1; FLJ35509; Heterogeneous nuclear ribonucleoprotein L; heterogenous nuclear ribonucleoprotein L; hnRNP L; Hnrnpl; hnRNP-L; Hnrpl; P/OKcl.14
Gene Symbols HNRNPL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 282-589 of human HNRNPL (NP_0015242).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15388, 3191, 80846
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.