Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP L Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | hnRNP L |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
hnRNP L Polyclonal specifically detects hnRNP L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
hnRNP L | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL | |
Recombinant fusion protein containing a sequence corresponding to amino acids 282-589 of human hnRNP L (NP_001524.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
PBS with 50% glycerol, pH7.3. | |
3191 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title