Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP LL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561145
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561145 Supplier Invitrogen™ Supplier No. PA561145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QASKNIIQPP SCVLHYYNVP LCVTEETFTK LCNDHEVLTF IKYKVFDAKP SAKTLSGLLE WECKTD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

HNRNPLL is a master regulator of activation-induced alternative splicing in T cells. In particular, it alters splicing of CD45 (PTPRC), a tyrosine phosphatase essential for T-cell development and activation (Oberdoerffer et al., 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP LL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hnrnpll
Gene Accession No. Q8WVV9
Gene Alias 2510028H02Rik; 2810036L13Rik; AI256697; AI852082; BLOCK24; heterogeneous nuclear ribonucleoprotein L like; heterogeneous nuclear ribonucleoprotein L-like; Hnrnpll; HNRPLL; RGD1305861; SRRF; stromal RNA regulating factor; stromal RNA-regulating factor
Gene Symbols Hnrnpll
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human hnRNP LL. Recombinant protein control fragment (Product #RP-96109).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.