Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hnRNP LL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587841
Description
Immunogen sequence: IRNDNDSWDY TKPYLGRRDR GKGRQRQAIL GEHPSSFRHD GYGSHGPLLP LPSRYRMGSR DTPELVAYPL PQASSSYMHG GNPSGSVVMV SGLHQLKMNC S; Positive Samples: A-549, DU145, HepG2, 293T, Mouse liver.
HNRNPLL is a master regulator of activation-induced alternative splicing in T cells. In particular, it alters splicing of CD45 (PTPRC), a tyrosine phosphatase essential for T-cell development and activation (Oberdoerffer et al., 2008).
Specifications
hnRNP LL | |
Polyclonal | |
Unconjugated | |
Hnrnpll | |
2510028H02Rik; 2810036L13Rik; AI256697; AI852082; BLOCK24; heterogeneous nuclear ribonucleoprotein L like; heterogeneous nuclear ribonucleoprotein L-like; Hnrnpll; HNRPLL; RGD1305861; SRRF; stromal RNA regulating factor; stromal RNA-regulating factor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
72692, 92906 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.68 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WVV9, Q921F4 | |
Hnrnpll | |
Recombinant fusion protein containing a sequence corresponding to amino acids 250-350 of human HNRPLL (NP_6124032). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction