Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hnRNP LL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106669
Description
Antibody detects endogenous levels of total HNRPLL.
HNRNPLL is a master regulator of activation-induced alternative splicing in T cells. In particular, it alters splicing of CD45 (PTPRC), a tyrosine phosphatase essential for T-cell development and activation (Oberdoerffer et al., 2008).
Specifications
| hnRNP LL | |
| Polyclonal | |
| Unconjugated | |
| Hnrnpll | |
| 2510028H02Rik; 2810036L13Rik; AI256697; AI852082; BLOCK24; heterogeneous nuclear ribonucleoprotein L like; heterogeneous nuclear ribonucleoprotein L-like; Hnrnpll; HNRPLL; RGD1305861; SRRF; stromal RNA regulating factor; stromal RNA-regulating factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 313842, 72692, 92906 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WVV9, Q921F4 | |
| Hnrnpll | |
| A synthesized peptide derived from human HNRNPLL(Accession Q8WVV9), corresponding to amino acid residues R243-S293. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction