Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP M Antibody (2A6), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

$206.50 - $499.50

Specifications

Antigen hnRNP M
Clone 2A6
Dilution Western Blot 1:1000, Simple Western 1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB200315SS
SDP
View Documents
Novus Biologicals
NB200315SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB200315
SDP
View Documents
Novus Biologicals
NB200315
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

hnRNP M Monoclonal specifically detects hnRNP M in Human, Mouse, Rat, Porcine, Bovine, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

hnRNP M
Western Blot 1:1000, Simple Western 1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Ascites
RUO
Human, Mouse, Rat, Pig, Bovine, Rabbit
P52272
4670
M19 fusion protein containing full-length human protein. [UniProt# P52272]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
68 kDa
2A6
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Cancer
Ascites with 0.1% Sodium Azide
DKFZp547H118, heterogeneous nuclear ribonucleoprotein M, hnRNP M, HNRNPM4, HNRPM4heterogenous nuclear ribonucleoprotein M, HNRPMheterogenous nuclear ribonucleoprotein M4, HTGR1, M4 protein, N-acetylglucosamine receptor 1, NAGR1hnRNA-binding protein M4
HNRNPM
IgG2b κ
Unpurified
NB200-315 is specific for hnRNP M3 and M4 proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.