Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP M1-M4 Antibody (1D8), Novus Biologicals™
SDP

Catalog No. p-5000874 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200314SS 0.025 mL
NB200314 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB200314SS Supplier Novus Biologicals Supplier No. NB200314SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

hnRNP M1-M4 Monoclonal specifically detects hnRNP M1-M4 in Human, Mouse, Rat, Porcine, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen hnRNP M1-M4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1D8
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:10-1:500
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. P52272
Gene Alias DKFZp547H118, heterogeneous nuclear ribonucleoprotein M, hnRNP M, HNRNPM4, HNRPM4heterogenous nuclear ribonucleoprotein M, HNRPMheterogenous nuclear ribonucleoprotein M4, HTGR1, M4 protein, N-acetylglucosamine receptor 1, NAGR1hnRNA-binding protein M4
Gene Symbols HNRNPM
Host Species Mouse
Immunogen M19 fusion protein containing full-length human protein. [UniProt# P52272]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4670
Test Specificity NB200-314 is specific for all four M proteins of hnRNP (M1, M2, M3 and M4).
Target Species Human, Mouse, Rat, Pig, Bovine, Rabbit
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.