Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hnRNP M1-M4 Monoclonal Antibody (1D8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA116673
Description
Suggested positive control: antigen standard for HNRNPM (transient overexpression lysate).
Proteins that directly bind to nascent RNA polymerase II transcripts, the heterogenous nuclear ribonucleoproteins (hnRNPs), play an important role in both transcript-specific packaging and alternative splicing of pre-mRNAs. A group of abundant hnRNPs, the M1-M4 proteins, appear as a cluster of four proteins. The M proteins are pre-mRNA binding proteins in vivo, and they bind avidly to poly (G) and poly (U) RNA homopolymers in vitro. M proteins are members of the ribonucleoprotein consensus sequence family of RNA-binding proteins with greatest similarity to a hypothetical RNA-binding protein from Saccharomyces cerevisiae. The M proteins also possess an unusual hexapeptide-repeat region rich in methionine and arginine residues (MR repeat motif) that resembles a repeat in the 64kDa subunit of cleavage stimulation factor, which is involved in 3'-end maturation of pre-mRNAs. Proteins immunologically related to M exist in divergent eukaryotes ranging from human to yeast.
Specifications
hnRNP M1-M4 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.1% sodium azide | |
P52272, Q62826, Q9D0E1 | |
HNRNPM | |
M19 fusion protein containing full-length human protein. | |
RUO | |
100356502, 100514153, 116655, 4670, 515290, 76936 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1D8 | |
Unconjugated | |
HNRNPM | |
2610023M21Rik; AA409009; CEA receptor; CEAR; Heterogeneous nuclear ribonucleoprotein M; heterogenous nuclear ribonucleoprotein M4; hnRNA-binding protein M4; hnRNP M; HNRNP3; Hnrnpm; HNRNPM3; HNRNPM4; HNRPM; Hnrpm4; HTGR1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein M; M4 protein; mKIAA4193; N-acetylglucosamine receptor 1; NAGR1 | |
Mouse | |
200 μL | |
Primary | |
Human, Mouse, Rat, Rabbit, Bovine, Pig | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction