Learn More
hnRNP M3/4 Mouse anti-Human, Mouse, Clone: 2A6-2H3, Bethyl Laboratories
Mouse Monoclonal Antibody
Supplier: Bethyl Laboratories, Inc A500011A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Proteins that directly bind to nascent RNA polymerase II transcripts, the heterogenous nuclear ribonucleoproteins (hnRNPs), play an important role in both transcript-specific packaging and alternative splicing of pre-mRNAs. A group of abundant hnRNPs, the M1-M4 proteins, appear as a cluster of four proteins. The M proteins are pre-mRNA binding proteins in vivo, and they bind avidly to poly (G) and poly (U) RNA homopolymers in vitro. M proteins are members of the ribonucleoprotein consensus sequence family of RNA-binding proteins with greatest similarity to a hypothetical RNA-binding protein from Saccharomyces cerevisiae. The M proteins also possess an unusual hexapeptide-repeat region rich in methionine and arginine residues (MR repeat motif) that resembles a repeat in the 64kDa subunit of cleavage stimulation factor, which is involved in 3′-end maturation of pre-mRNAs. Proteins immunologically related to M exist in divergent eukaryotes ranging from human to yeast.Specifications
hnRNP M3/4 | |
Monoclonal | |
Unconjugated | |
HNRNPM | |
heterogeneous nuclear ribonucleoprotein M, HNRNPM, HNRNPM4, HNRPM, hnRNA-binding protein M4, HNRNPM3, hnRNA-binding protein M3, HNRNP3, N-acetylglucosamine receptor 1, M4 protein | |
Mouse | |
Affinity chromatography | |
RUO | |
4670, 76936 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
2A6-2H3 | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P52272, Q9D0E1 | |
Hnrnpm | |
hnRNP M1-4 proteins | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.