Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP M3/4 Mouse anti-Human, Mouse, Clone: 2A6-2H3, Bethyl Laboratories

Catalog No. 501571232
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1232 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-1232 Supplier Bethyl Laboratories, Inc Supplier No. A500011AT
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Proteins that directly bind to nascent RNA polymerase II transcripts, the heterogenous nuclear ribonucleoproteins (hnRNPs), play an important role in both transcript-specific packaging and alternative splicing of pre-mRNAs. A group of abundant hnRNPs, the M1-M4 proteins, appear as a cluster of four proteins. The M proteins are pre-mRNA binding proteins in vivo, and they bind avidly to poly (G) and poly (U) RNA homopolymers in vitro. M proteins are members of the ribonucleoprotein consensus sequence family of RNA-binding proteins with greatest similarity to a hypothetical RNA-binding protein from Saccharomyces cerevisiae. The M proteins also possess an unusual hexapeptide-repeat region rich in methionine and arginine residues (MR repeat motif) that resembles a repeat in the 64kDa subunit of cleavage stimulation factor, which is involved in 3′-end maturation of pre-mRNAs. Proteins immunologically related to M exist in divergent eukaryotes ranging from human to yeast.

Specifications

Antigen hnRNP M3/4
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2A6-2H3
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene HNRNPM
Gene Accession No. P52272, Q9D0E1
Gene Alias heterogeneous nuclear ribonucleoprotein M, HNRNPM, HNRNPM4, HNRPM, hnRNA-binding protein M4, HNRNPM3, hnRNA-binding protein M3, HNRNP3, N-acetylglucosamine receptor 1, M4 protein
Gene Symbols HNRNPM
Host Species Mouse
Immunogen hnRNP M1-4 proteins
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4670, 76936
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.