Learn More
Invitrogen™ hnRNP M3-M4 Monoclonal Antibody (2A6)

Description
Suggested positive control: antigen standard for HNRNPM (transient overexpression lysate).
Specifications
Specifications
| Antigen | hnRNP M3-M4 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2A6 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.1% sodium azide |
| Gene | Hnrnpm |
| Gene Accession No. | P52272, Q62826, Q9D0E1 |
| Gene Alias | 2610023M21Rik; AA409009; CEA receptor; CEAR; heterogeneous nuclear ribonucleoprotein M; heterogenous nuclear ribonucleoprotein M4; hnRNA-binding protein M4; hnRNP M; HNRNP3; HNRNPM; HNRNPM3; HNRNPM4; HNRPM; HNRPM4; HTGR1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein M; M4 protein; mKIAA4193; N-acetylglucosamine receptor 1; NAGR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.