Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HnRNP Q Recombinant Rabbit Monoclonal Antibody (JG35-72)
GREENER_CHOICE

Catalog No. PIMA534735
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534735 Supplier Invitrogen™ Supplier No. MA534735
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: K562, LOVO, SiHa, rat testis tissue, human thyroid gland tissue, human breast tissue, human kidney tissue.

Heterogenous nuclear ribonucleoprotein (hnRNP) implicated in mRNA processing mechanisms. Component of the CRD-mediated complex that promotes MYC mRNA stability. Isoform 1, isoform 2 and isoform 3 are associated in vitro with pre-mRNA, splicing intermediates and mature mRNA protein complexes. Isoform 1 binds to apoB mRNA AU-rich sequences. Isoform 1 is part of the APOB mRNA editosome complex and may modulate the postranscriptional C to U RNA-editing of the APOB mRNA through either by binding to A1CF (APOBEC1 complementation factor), to APOBEC1 or to RNA itself. May be involved in translationally coupled mRNA turnover. Implicated with other RNA-binding proteins in the cytoplasmic deadenylation/translational and decay interplay of the FOS mRNA mediated by the major coding-region determinant of instability (mCRD) domain. Interacts in vitro preferentially with poly(A) and poly(U) RNA sequences. Isoform 3 may be involved in cytoplasmic vesicle-based mRNA transport through interaction with synaptotagmins. Component of the GAIT (gamma interferon-activated inhibitor of translation) complex which mediates interferon-gamma-induced transcript-selective translation inhibition in inflammation processes. Upon interferon-gamma activation assembles into the GAIT complex which binds to stem loop-containing GAIT elements in the 3'-UTR of diverse inflammatory mRNAs (such as ceruplasmin) and suppresses their translation; seems not to be essential for GAIT complex function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HnRNP Q
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JG35-72
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Syncrip
Gene Accession No. O60506
Gene Alias 2610109K23Rik; 4632417O19Rik; Ab2-339; Glycine- and tyrosine-rich RNA-binding protein; GRYRBP; GRY-RBP; heterogeneous nuclear ribonucleoprotein Q; hnRNP; hnRNP Q; HNRNPQ; hnRNP-Q; HNRPQ; HNRPQ1; liver regeneration-related protein LRRG077; NS1-associated protein 1; NSAP1; Nsap1l; PP68; RRM RNA binding protein GRY-RBP; synaptotagmin binding cytoplasmic RNA interacting protein; synaptotagmin binding, cytoplasmic RNA interacting protein; synaptotagmin-binding, cytoplasmic RNA-interacting protein; Syncrip
Gene Symbols Syncrip
Host Species Rabbit
Immunogen Synthetic peptide within Human hnRNP Q aa 574-623.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10492
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.