Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hnRNP R Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528473
Description
Recommended positive controls: 293T, A431, HeLa. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (81%), Chicken (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs and also contains a nuclear localization motif. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
hnRNP R | |
Polyclonal | |
Unconjugated | |
HNRNPR | |
2610003J05Rik; 2610528B01Rik; FLJ25714; heterogeneous nuclear ribonucleoprotein R; hnRNP R; HNRNPR; hnRNP-R; HNRPR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10236 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O43390 | |
HNRNPR | |
Recombinant fragment corresponding to a region within amino acids 1 and 187 of Human hnRNP R. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction