Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP U Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002135AF405
Description
hnRNP U Polyclonal antibody specifically detects hnRNP U in Human, Mouse samples. It is validated for Western BlotSpecifications
hnRNP U | |
Polyclonal | |
Western Blot | |
heterogeneous nuclear ribonucleoprotein U, heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A), hnRNP U, HNRPU, p120, p120 nuclear protein, pp120, SAFA, SAF-AhnRNPU, Scaffold attachment factor A, U21.1 | |
The immunogen recognized by this antibody maps to a region between residue 775 and the C-terminus (residue 824) of human Heterogeneous Nuclear Ribonucleoprotein U using the numbering given in entry NP_114032.1 (GeneID 3192). | |
0.1 mL | |
Cellular Markers | |
3192 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction