Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP-U Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | hnRNP-U |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-8227
|
Bethyl Laboratories, Inc
A300690AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
| hnRNP-U | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| C1orf199, chromosome 1 open reading frame 199, FLJ30202, FLJ37978, heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A), hnRNP U, HNRNPU antisense RNA 1, HNRNPU antisense RNA 1 (non-protein coding), HNRNPU-AS1, HNRPU, NCRNA00201, non-protein coding RNA 201, p120, p120 nuclear protein, pp120, SAFA, SAF-A, Scaffold attachment factor A, U21.1 | |
| Between 775 and C-term | |
| Primary | |
| 4° C | |
| HNRNPU |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q00839, Q8VEK3 | |
| 3192, 51810 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title