Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP U Rabbit anti-Human, Mouse, Rat, Clone: JB23-43, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275526
Description
hnRNP U Monoclonal specifically detects hnRNP U in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| hnRNP U | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:400 | |
| heterogeneous nuclear ribonucleoprotein U, heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A), hnRNP U, HNRPU, p120, p120 nuclear protein, pp120, SAFA, SAF-AhnRNPU, Scaffold attachment factor A, U21.1 | |
| Rabbit | |
| 120 kDa | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| JB23-43 | |
| Unconjugated | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| HNRNPU | |
| Recombinantpeptide within Human hnRNP U aa 1-140/ 825. (SwissProt: Q00839 Human; SwissProt: Q8VEK3 Mouse; SwissProt: Q6IMY8 Rat) | |
| Protein A purified | |
| Cellular Markers | |
| 3192.0 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction