Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP UL1 Monoclonal Antibody (EAP 4A11), eBioscience™, Invitrogen™

Catalog No. 14980637
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-980-637 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-980-637 Supplier Invitrogen™ Supplier No. 14980637
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: This EAP 4A11 monoclonal antibody recognizes human hnRNP UL1. EAP 4A11 detects a ∽95 kDa band corresponding to hnRNP UL1 by Western Blot and stains hnRNP UL1 in the nucleus in Immunocytochemistry. Applications Reported: This EAP 4A11 antibody has been reported for use in western blot and immunocytochemistry/immunofluorescence. Applications Tested: This EAP 4A11 antibody has been tested by western blot of human cell lysates and immunocytochemistry/immunofluorescence of human cell lines. This may be used at less than or equal to 4.0 μg/ml for western blot and less than or equal to 2.5 μg/mL for immunocytochemistry/immunofluorescence.

HNRPUL1 is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells.This gene encodes a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells. Four transcript variants encoding different isoforms have been found for this gene. Another variant has also been found, but its full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP UL1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone EAP 4A11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Hnrnpul1
Gene Accession No. Q9BUJ2
Gene Alias Adenovirus early region 1B-associated protein 5; E130317O14Rik; E1B 55kDa associated protein 5; E1B-55 kDa-associated protein 5; E1BAP5; E1B-AP5; EIB-55kDa associated protein 5; heterogeneous nuclear ribonucleoprotein U like 1; heterogeneous nuclear ribonucleoprotein U-like 1; heterogeneous nuclear ribonucleoprotein U-like protein 1; Hnrnpul; Hnrnpul1; HNRPUL1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein U-like protein 1
Gene Symbols Hnrnpul1
Host Species Rat
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11100
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.