Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP UL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566947 Supplier Invitrogen™ Supplier No. PA566947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQYNRDWQSYY YHHPQDRDRY YRNYYGYQGY R Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

HNRNPUL2 gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP UL2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HNRNPUL2
Gene Accession No. Q1KMD3
Gene Alias 1110031M08Rik; AI465155; heterogeneous nuclear ribonucleoprotein U like 2; heterogeneous nuclear ribonucleoprotein U-like 2; heterogeneous nuclear ribonucleoprotein U-like protein 2; Hnrnpul2; Hnrpul2; Manp; MLF1-associated nuclear protein; SAF-A2; Scaffold-attachment factor A2
Gene Symbols HNRNPUL2
Host Species Rabbit
Immunogen Recombinant Human hnRNP UL2. Recombinant protein control fragment (Product #RP-105434).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.