Learn More
Invitrogen™ HNRNPA0 Polyclonal Antibody

Description
Immunogen sequence: KLFVGGLKGDV AEGDLIEHFS QFGTVE Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
Specifications
Specifications
| Antigen | HNRNPA0 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Hnrnpa0 |
| Gene Accession No. | Q13151 |
| Gene Alias | 1110055B05Rik; 3010025E17Rik; Heterogeneous nuclear ribonucleoprotein A0; hnRNA binding protein; hnRNP A0; HNRNPA0; Hnrpa0 |
| Gene Symbols | Hnrnpa0 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.