Learn More
HNRNPF Mouse anti-Human, Mouse, Rat, Clone: 2E7A4, ProteinTech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 677011IG150UL

Description
Aliquoting is unnecessary for -20°C storage.
HNRNPF Monoclonal antibody specifically detects HNRNPF in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
HNRNPF | |
Monoclonal | |
500 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P52597, Q794E4, Q9Z2X1 | |
Hnrnpf | |
HNRNPF Fusion Protein Ag7128 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
2E7A4 | |
Unconjugated | |
Hnrnpf | |
4833420I20Rik; AA407306; heterogeneous nuclear ribonucleoprotein F; Heterogeneous nuclear ribonucleoprotein F, N-terminally processed; hnRNP F; HnRNP F protein; hnRNP H; Hnrnpf; HNRNPH1; Hnrpf; mcs94 1; mcs94-1; Nucleolin-like protein mcs94-1; OK/SW-cl.23; ribonucleoprotein F; ROF | |
Mouse | |
Protein G | |
RUO | |
3185, 64200, 98758 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.