Learn More
HNRNPL Mouse anti-Human, Clone: OTI9G7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805445

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Heterogeneous nuclear RNAs which include mRNA precursors and mature mRNAs are associated with specific proteins to form heterogenous ribonucleoprotein complexes. Heterogeneous nuclear ribonucleoprotein L is among the proteins that are stably associated with hnRNP complexes and along with other hnRNP proteins is likely to play a major role in the formation, packaging, processing, and function of mRNA. Heterogeneous nuclear ribonucleoprotein L is present in the nucleoplasm as part of the HNRP complex. HNRP proteins have also been identified outside of the nucleoplasm. Exchange of hnRNP for mRNA-binding proteins accompanies transport of mRNA from the nucleus to the cytoplasm. Since HNRP proteins have been shown to shuttle between the nucleus and the cytoplasm, it is possible that they also have cytoplasmic functions. Two transcript variants encoding different isoforms have been found for this gene.Specifications
HNRNPL | |
Monoclonal | |
Unconjugated | |
HNRNPL | |
HNRPL, P/OKcl.14, hnRNP-L | |
Mouse | |
Affinity Chromatography | |
RUO | |
3191 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI9G7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P14866 | |
HNRNPL | |
Human recombit protein fragment corresponding to amino acids 270-502 of human HNRNPL produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.