Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HNRNPR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109826
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109826 Supplier Invitrogen™ Supplier No. PA5109826
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGRGGGRGGR GAPPPPRGRG APPPRGRAGY SQRGAPLGPP RGSRGGRGGP AQQQRGRGSR GSRGNRGGNV GGKRKADGYN QPDSKRRQTN NQQNWGSQPI AQQPLQQGGD YSGNYGYNND NQEFYQDTYG QQWK.

This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs and also contains a nuclear localization motif. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNRNPR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HNRNPR
Gene Accession No. O43390
Gene Alias 2610003J05Rik; 2610528B01Rik; FLJ25714; heterogeneous nuclear ribonucleoprotein R; hnRNP R; HNRNPR; hnRNP-R; HNRPR
Gene Symbols HNRNPR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human HNRNPR (NP_005817.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10236, 319110, 74326
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.