Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNRNPUL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | HNRNPUL1 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
HNRNPUL1 Polyclonal specifically detects HNRNPUL1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HNRNPUL1 | |
Polyclonal | |
Rabbit | |
Human | |
Adenovirus early region 1B-associated protein 5, E1B-AP5E1B 55kDa associated protein 5, E1BAP5E1B-55 kDa-associated protein 5, FLJ12944, heterogeneous nuclear ribonucleoprotein U-like 1, heterogeneous nuclear ribonucleoprotein U-like protein 1, HNRPUL1E1B-55kDa-associated protein 5 | |
HNRNPUL1 | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
11100 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PTAQTYPQPSYNQYQQYAQQWNQYYQNQGQWPPYYGNYDYGSYSGNTQGGTSTQ | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title