Learn More
HNRNPUL2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304619A

Description
The recommended shelf life for this product is 1 year from date of receipt.
HNRNPUL2 gene ontology annotations related to this gene include RNA binding.Specifications
HNRNPUL2 | |
Polyclonal | |
Unconjugated | |
HNRNPUL2 | |
HNRPUL2, SAF-A2, heterogeneous nuclear ribonucleoprotein U-like protein 2, scaffold-attachment factor A2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221092, 68693 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q00PI9, Q1KMD3 | |
Hnrnpul2 | |
Between 25 and 75 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.